siRNA Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182981.2 | NP_892026.1 | 6 | 1195 | ||
| GenBank mRNA | AF191740.1 | - | 4 | 833 | ||
| AK290383.1 | - | 6 | 1177 | |||
| AY037158.1 | - | 6 | 1160 | |||
| AY258066.1 | - | 7 | 1646 | |||
| AY258067.1 | - | 6 | 1486 | |||
| AY258068.1 | - | 6 | 1176 | |||
| BC020434.1 | - | 6 | 1161 | |||
| BC032476.1 | - | 6 | 1160 | 5215262 | ||
| BC093687.1 | - | 6 | 1078 | 7939532 | ||
| BC113417.1 | - | 6 | 1078 | 8322469 | ||

OSGIN1
Genomic Map