siRNA Details
| Gene Symbol: MRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013393.1 | NP_037525.1 | 3 | 479 | ||
| GenBank mRNA | AF093415.1 | - | 3 | 479 | ||
| AK024635.1 | - | 2 | 228 | |||
| AK302060.1 | - | 4 | 590 | |||
| BC017106.2 | - | 2 | 442 | |||
| BC114514.1 | - | 3 | 464 | 40031660 | ||
| BC114564.1 | - | 3 | 433 | |||
| CR749618.1 | - | 3 | 488 | |||
| EU794658.1 | - | 3 | 479 | |||

MRM2
Genomic Map