siRNA Details
| Gene Symbol: PRICKLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013397.5 | NP_037529.3 | 7 | 877 | ||
| GenBank mRNA | BC110458.1 | - | 5 | 1324 | ||
| BC110459.1 | - | 5 | 1329 | 40024767 | ||
| DA917216.1 | - | 3 | 491 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRICKLE4| Gene Symbol: PRICKLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013397.5 | NP_037529.3 | 7 | 877 | ||
| GenBank mRNA | BC110458.1 | - | 5 | 1324 | ||
| BC110459.1 | - | 5 | 1329 | 40024767 | ||
| DA917216.1 | - | 3 | 491 | |||