siRNA Details
| Gene Symbol: HBM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003938.3 | NP_001003938.1 | 2 | 150 | ||
| GenBank mRNA | AY698022.2 | - | 2 | 150 | ||
| BC035682.1 | - | 2 | 133 | 5749846 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HBM| Gene Symbol: HBM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003938.3 | NP_001003938.1 | 2 | 150 | ||
| GenBank mRNA | AY698022.2 | - | 2 | 150 | ||
| BC035682.1 | - | 2 | 133 | 5749846 | ||