siRNA Details
| Gene Symbol: EHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014601.3 | NP_055416.2 | 5, 6 | 1341 | ||
| GenBank mRNA | AF181263.1 | - | 4, 5 | 1129 | ||
| AF454952.1 | - | 4, 5 | 1146 | |||
| AK074701.1 | - | 5, 6 | 1263 | |||
| AK097126.1 | - | 5, 6 | 668 | |||
| AK097228.1 | - | 4, 5 | 660 | |||
| AK293429.1 | - | 1, 2 | 190 | |||
| AK296907.1 | - | 6, 7 | 1233 | |||
| AK298067.1 | - | 4, 5 | 804 | |||
| AK300512.1 | - | 2, 3 | 292 | |||
| AK315548.1 | - | 5, 6 | 1265 | |||
| AK316448.1 | - | 2, 3 | 241 | |||

EHD2
Genomic Map