siRNA Details
| Gene Symbol: ONECUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004498.2 | NP_004489.1 | 2 | 1314 | ||
| NR_073510.1 | 2 | 370 | ||||
| GenBank mRNA | BC140830.1 | - | 2 | 1281 | 9021698 | |
| BP289459.1 | - | 2 | 131 | |||
| BX118406.1 | - | 2 | 370 | |||
| EU532019.1 | - | 2 | 934 | |||
| U77975.1 | - | 2 | 667 | |||
| U96173.1 | - | 2 | 1314 | |||

ONECUT1
Genomic Map