siRNA Details
| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131065.1 | NP_001124537.1 | 4 | 1638 | ||
| NM_001131066.1 | NP_001124538.1 | 4 | 483 | |||
| NM_173362.3 | NP_775498.1 | 3 | 1540 | |||
| XM_011543360.2 | XP_011541662.1 | 5 | 1526 | |||
| XM_011543361.2 | XP_011541663.1 | 3 | 418 | |||
| XM_017009413.1 | XP_016864902.1 | 4 | 650 | |||
| GenBank mRNA | BC024023.1 | - | 2 | 257 | 4670524 | |
| BC035110.2 | - | 3 | 1538 | 5262922 | ||
| BC048979.1 | - | 3 | 1565 | 4838006 | ||
| BI551847.1 | - | 4 | 486 | |||
| BM263054.1 | - | 3 | 447 | |||

RFESD
Genomic Map