siRNA Details
| Gene Symbol: KLHL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152467.3 | NP_689680.2 | 1 | 243 | ||
| XM_011524706.2 | XP_011523008.1 | 3 | 376 | |||
| GenBank mRNA | AK057224.1 | - | 1 | 206 | ||
| AK302141.1 | - | 1 | 209 | |||
| AY495339.1 | - | 1 | 225 | |||
| BC067753.1 | - | 1 | 230 | |||
| BP371285.1 | - | 1 | 243 | |||

KLHL10
Genomic Map