siRNA Details
| Gene Symbol: TLX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016170.4 | NP_057254.1 | 3 | 1040 | ||
| GenBank mRNA | AB008501.1 | - | 3 | 702 | ||
| AB593086.1 | - | 4 | 727 | |||
| BC006356.2 | - | 3 | 1025 | 4125397 | ||
| BC006396.1 | - | 2 | 1297 | |||
| BC018611.1 | - | 4 | 636 | |||

TLX2
Genomic Map