siRNA Details
| Gene Symbol: HOXA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002141.4 | NP_002132.3 | 2 | 889 | ||
| GenBank mRNA | BC126994.1 | - | 2 | 289 | ||
| M74297.1 | - | 2 | 842 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXA4| Gene Symbol: HOXA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002141.4 | NP_002132.3 | 2 | 889 | ||
| GenBank mRNA | BC126994.1 | - | 2 | 289 | ||
| M74297.1 | - | 2 | 842 | |||