siRNA Details
| Gene Symbol: HOXA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024014.3 | NP_076919.1 | 1 | 420 | ||
| GenBank mRNA | BC069497.1 | - | 1 | 395 | 7262260 | |
| BC104915.1 | - | 1 | 420 | 8143918 | ||
| BC104917.1 | - | 1 | 420 | 8143920 | ||
| BG749730.1 | - | 1 | 164 | |||
| Gene Symbol: HOXA-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038831.1 | 1 | 189 | |||
| GenBank mRNA | AK091933.1 | - | 1 | 189 | ||

HOXA6
Genomic Map