siRNA Details
| Gene Symbol: SNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178310.3 | NP_840101.1 | 2 | 392 | ||
| GenBank mRNA | BC041461.1 | - | 2 | 299 | ||
| BC107058.2 | - | 2 | 392 | 40005947 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNAI3| Gene Symbol: SNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178310.3 | NP_840101.1 | 2 | 392 | ||
| GenBank mRNA | BC041461.1 | - | 2 | 299 | ||
| BC107058.2 | - | 2 | 392 | 40005947 | ||