siRNA Details
| Gene Symbol: KRTAP13-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181622.1 | NP_853653.1 | 1 | 195 | ||
| GenBank mRNA | AB096940.1 | - | 1 | 209 | ||
| BC103965.2 | - | 1 | 195 | 40014483 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP13-3| Gene Symbol: KRTAP13-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181622.1 | NP_853653.1 | 1 | 195 | ||
| GenBank mRNA | AB096940.1 | - | 1 | 209 | ||
| BC103965.2 | - | 1 | 195 | 40014483 | ||