siRNA Details
| Gene Symbol: TMEM119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181724.2 | NP_859075.2 | 2 | 486 | ||
| XM_011538271.2 | XP_011536573.1 | 3 | 676 | |||
| GenBank mRNA | AK075501.1 | - | 2 | 376 | ||
| AK295288.1 | - | 3 | 303 | |||
| AY358389.1 | - | 2 | 485 | |||
| BC096825.1 | - | 2 | 397 | 30558500 | ||

TMEM119
Genomic Map