siRNA Details
| Gene Symbol: TMEM119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181724.2 | NP_859075.2 | 2 | 531 | ||
| XM_011538271.2 | XP_011536573.1 | 3 | 721 | |||
| GenBank mRNA | AK075501.1 | - | 2 | 421 | ||
| AL521683.3 | - | 2 | 536 | |||
| AY358389.1 | - | 2 | 530 | |||
| BC096825.1 | - | 2 | 442 | 30558500 | ||

TMEM119
Genomic Map