siRNA Details
| Gene Symbol: CPLX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181654.3 | NP_857637.1 | 1 | 230 | ||
| GenBank mRNA | AB593158.1 | - | 1 | 211 | ||
| AK123184.1 | - | 1 | 230 | |||
| AY286502.1 | - | 1 | 60 | |||
| BC106886.2 | - | 3 | 635 | |||
| BC106887.1 | - | 1 | 223 | 40030305 | ||
| BC106888.1 | - | 1 | 223 | 40030309 | ||
| BX640655.1 | - | 1 | 209 | |||
| BX648518.1 | - | 1 | 213 | |||

CPLX4
Genomic Map