siRNA Details
| Gene Symbol: LINGO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004432.3 | NP_001004432.1 | 2 | 1717 | ||
| GenBank mRNA | AY358115.1 | - | 2 | 1684 | ||
| BC137220.1 | - | 2 | 1541 | 9021217 | ||
| BC137221.1 | - | 2 | 1541 | 9021218 | ||

LINGO4
Genomic Map