siRNA Details
| Gene Symbol: ZBTB8OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308135.1 | NP_001295064.1 | 1 | 38 | ||
| NM_001308136.1 | NP_001295065.1 | 1 | 38 | |||
| NM_001308137.1 | NP_001295066.1 | 1 | 38 | |||
| NM_001308138.1 | NP_001295067.1 | 1 | 38 | |||
| NM_001308139.1 | NP_001295068.1 | 1 | 38 | |||
| NM_001308140.1 | NP_001295069.1 | 1 | 38 | |||
| NM_001308141.1 | NP_001295070.1 | 1 | 38 | |||
| NM_178547.3 | NP_848642.1 | 1 | 38 | |||
| XM_011541327.2 | XP_011539629.1 | 1 | 461 | |||
| XM_011541330.2 | XP_011539632.1 | 1 | 443 | |||
| XM_011541334.2 | XP_011539636.1 | 1 | 395 | |||
| XM_011541338.2 | XP_011539640.1 | 1 | 434 | |||
| XM_017001135.1 | XP_016856624.1 | 1 | 461 | |||
| XM_017001136.1 | XP_016856625.1 | 1 | 461 | |||
| GenBank mRNA | AK289480.1 | - | 1 | 380 | ||
| AY099493.1 | - | 1 | 32 | |||
| AY151084.1 | - | 1 | 32 | |||
| AY151085.1 | - | 1 | 32 | |||
| AY158005.1 | - | 1 | 13 | |||
| CK819207.1 | - | 5 | 530 | |||

ZBTB8OS
Genomic Map