siRNA Details
| Gene Symbol: SLC35E4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001479.3 | NP_001001479.1 | 2 | 1426 | ||
| XM_017028794.1 | XP_016884283.1 | 3 | 2518 | |||
| XM_017028795.1 | XP_016884284.1 | 3 | 2092 | |||
| GenBank mRNA | BC040191.1 | - | 2 | 1434 | 4814106 | |
| BC107119.2 | - | 2 | 812 | 40023535 | ||
| CR456351.1 | - | 2 | 915 | |||

SLC35E4
Genomic Map