siRNA Details
| Gene Symbol: FAM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182761.3 | NP_877438.2 | 2, 3 | 421 | ||
| GenBank mRNA | BC040622.1 | - | 2, 3 | 382 | ||
| BC128242.1 | - | 2, 3 | 294 | |||
| BC128243.1 | - | 2, 3 | 294 | 40114744 | ||

FAM170A
Genomic Map