siRNA Details
| Gene Symbol: RSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317942.1 | NP_001304871.1 | 2 | 158 | ||
| NM_178565.4 | NP_848660.3 | 3 | 837 | |||
| XM_011517018.1 | XP_011515320.1 | 3 | 853 | |||
| XM_011517019.1 | XP_011515321.1 | 3 | 853 | |||
| XM_017013395.1 | XP_016868884.1 | 3 | 826 | |||
| GenBank mRNA | AK123023.1 | - | 3 | 800 | ||
| AY358166.1 | - | 3 | 490 | |||
| BC027938.1 | - | 2 | 158 | 5201681 | ||

RSPO2
Genomic Map