siRNA Details
| Gene Symbol: CXorf67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203407.2 | NP_981952.1 | 1 | 616 | ||
| GenBank mRNA | BC046248.1 | - | 1 | 616 | 6046539 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXorf67| Gene Symbol: CXorf67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203407.2 | NP_981952.1 | 1 | 616 | ||
| GenBank mRNA | BC046248.1 | - | 1 | 616 | 6046539 | |