siRNA Details
| Gene Symbol: PALM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145028.1 | NP_001138500.1 | 3 | 254 | ||
| XM_005259891.4 | XP_005259948.1 | 4 | 455 | |||
| XM_011527965.2 | XP_011526267.1 | 3 | 375 | |||
| GenBank mRNA | AW130597.1 | - | 1 | 232 | ||
| CR747190.1 | - | 2 | 377 | |||

PALM3
Genomic Map