siRNA Details
| Gene Symbol: IFIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001547.4 | NP_001538.4 | 2 | 973 | ||
| GenBank mRNA | AA143609.1 | - | 1 | 422 | ||
| AK300431.1 | - | 2 | 875 | |||
| AK312831.1 | - | 2 | 922 | |||
| BC005987.1 | - | 2 | 876 | |||
| BC032839.2 | - | 2 | 876 | 4838844 | ||
| EB387611.1 | - | 1 | 349 | |||

IFIT2
Genomic Map