siRNA Details
Gene Symbol: GPR149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038705.1 | NP_001033794.1 | 2 | 1215 | ||
GenBank mRNA | AY255534.1 | - | 2 | 650 | ||
DQ318360.1 | - | 2 | 1215 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
