siRNA Details
| Gene Symbol: ZNF879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136116.1 | NP_001129588.1 | 5 | 589 | ||
| XM_005265908.3 | XP_005265965.1 | 5 | 1283 | |||
| XM_011534550.2 | XP_011532852.1 | 5 | 618 | |||
| XM_011534551.2 | XP_011532853.1 | 5 | 734 | |||
| GenBank mRNA | AK300504.1 | - | 5 | 521 | ||
| BX648737.1 | - | 5 | 559 | |||

ZNF879
Genomic Map