siRNA Details
| Gene Symbol: APOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000040.1 | NP_000031.1 | 4 | 247 | ||
| GenBank mRNA | BC027977.1 | - | 4 | 245 | 5229117 | |
| BC121081.1 | - | 4 | 247 | 40119733 | ||
| BC134419.1 | - | 4 | 247 | 40119734 | ||
| M28613.1 | - | 3 | 231 | |||
| M28614.1 | - | 3 | 231 | |||
| X00567.1 | - | 3 | 201 | |||
| X01388.1 | - | 4 | 272 | |||

APOC3
Genomic Map