siRNA Details
| Gene Symbol: IFNGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005534.3 | NP_005525.2 | 7 | 1556 | ||
| XM_005260969.2 | XP_005261026.1 | 8 | 1072 | |||
| XM_011529553.1 | XP_011527855.1 | 8 | 998 | |||
| XM_011529554.2 | XP_011527856.1 | 7 | 1432 | |||
| GenBank mRNA | AI824138.1 | - | 1 | 676 | ||
| AK292246.1 | - | 7 | 1004 | |||
| AK314005.1 | - | 7 | 1017 | |||
| BC003624.2 | - | 7 | 1024 | 2967074 | ||
| BQ008968.1 | - | 1 | 679 | |||
| U05875.1 | - | 7 | 1556 | |||
| U05877.1 | - | 7 | 1021 | |||

IFNGR2
Genomic Map