siRNA Details
| Gene Symbol: ITIH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198510.2 | NP_940912.1 | 8 | 2027 | ||
| GenBank mRNA | AY358170.1 | - | 8 | 2027 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITIH6| Gene Symbol: ITIH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198510.2 | NP_940912.1 | 8 | 2027 | ||
| GenBank mRNA | AY358170.1 | - | 8 | 2027 | ||