siRNA Details
| Gene Symbol: MCRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093767.2 | NP_001087236.1 | 4 | 305 | ||
| NM_001288798.1 | NP_001275727.1 | 5 | 441 | |||
| NM_207368.4 | NP_997251.2 | 4 | 305 | |||
| XM_011523571.2 | XP_011521873.1 | 4 | 378 | |||
| GenBank mRNA | AK000852.1 | - | 4 | 288 | ||
| AK125584.1 | - | 3 | 2357 | |||
| BC094876.1 | - | 5 | 415 | |||
| BG830426.1 | - | 4 | 305 | |||
| BX649156.1 | - | 4 | 303 | |||

MCRIP1
Genomic Map