siRNA Details
| Gene Symbol: PLET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145024.1 | NP_001138496.1 | 2 | 580 | ||
| GenBank mRNA | AY364431.1 | - | 2 | 309 | ||
| AY364434.1 | - | 1 | 126 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLET1| Gene Symbol: PLET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145024.1 | NP_001138496.1 | 2 | 580 | ||
| GenBank mRNA | AY364431.1 | - | 2 | 309 | ||
| AY364434.1 | - | 1 | 126 | |||