siRNA Details
| Gene Symbol: DPPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199286.3 | NP_954980.1 | 2, 3 | 453 | ||
| GenBank mRNA | AY317075.1 | - | 2, 3 | 453 | ||
| Gene Symbol: DPPA3P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY230136.1 | - | 1 | 392 | ||
| BC062480.1 | - | 1 | 585 | 30398001 | ||
| BC125145.1 | - | 1 | 404 | 40120129 | ||

DPPA3
Genomic Map