siRNA Details
| Gene Symbol: ZBTB41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194314.2 | NP_919290.2 | 10 | 2662 | ||
| NR_135153.1 | 11 | 2746 | ||||
| GenBank mRNA | AK128549.1 | - | 11 | 2725 | ||
| AW856335.1 | - | 1 | 365 | |||
| AY336933.1 | - | 7 | 1133 | |||
| BC132818.1 | - | 10 | 2626 | 40146840 | ||
| BC136600.1 | - | 10 | 2626 | 9020590 | ||

ZBTB41
Genomic Map