siRNA Details
| Gene Symbol: INSRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014215.2 | NP_055030.1 | 20 | 3793 | ||
| GenBank mRNA | J05046.1 | - | 19 | 3409 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

INSRR| Gene Symbol: INSRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014215.2 | NP_055030.1 | 20 | 3793 | ||
| GenBank mRNA | J05046.1 | - | 19 | 3409 | ||