siRNA Details
| Gene Symbol: IRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002199.3 | NP_002190.2 | 2 | 247 | ||
| GenBank mRNA | AA361545.1 | - | 2 | 183 | ||
| AK312953.1 | - | 2 | 225 | |||
| BC015803.2 | - | 1 | 54 | 3920890 | ||
| BP200998.1 | - | 2 | 247 | |||
| BT007264.1 | - | 1 | 47 | |||
| CR457077.1 | - | 1 | 47 | |||
| X15949.1 | - | 2 | 145 | |||

IRF2
Genomic Map