siRNA Details
| Gene Symbol: ITGA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000885.5 | NP_000876.3 | 14, 15 | 2292 | ||
| GenBank mRNA | AB208885.1 | - | 11, 12 | 1070 | ||
| BC016671.2 | - | 14, 15 | 1757 | |||
| L12002.1 | - | 14, 15 | 1957 | |||
| X16983.1 | - | 14, 15 | 1571 | |||

ITGA4
Genomic Map