siRNA Details
| Gene Symbol: CDRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006382.3 | NP_006373.2 | 12 | 2260 | ||
| GenBank mRNA | BC143291.1 | - | 5 | 536 | ||
| BC171723.1 | - | 3 | 298 | |||
| BU738380.1 | - | 1 | 517 | |||
| U43383.1 | - | 5 | 554 | |||
| U65652.1 | - | 5 | 789 | |||

CDRT1
Genomic Map