siRNA Details
| Gene Symbol: DKFZp779M0652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027134.1 | 1 | 215 | |||
| GenBank mRNA | BC126374.1 | - | 1 | 145 | ||
| BC126376.1 | - | 1 | 145 | |||
| BX640618.1 | - | 1 | 215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DKFZp779M0652| Gene Symbol: DKFZp779M0652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027134.1 | 1 | 215 | |||
| GenBank mRNA | BC126374.1 | - | 1 | 145 | ||
| BC126376.1 | - | 1 | 145 | |||
| BX640618.1 | - | 1 | 215 | |||