siRNA Details
| Gene Symbol: DNAJB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153614.3 | NP_705842.2 | 2 | 863 | ||
| XM_005273984.3 | XP_005274041.1 | 2 | 390 | |||
| XM_011545004.2 | XP_011543306.1 | 4 | 1114 | |||
| XM_011545005.2 | XP_011543307.1 | 4 | 1116 | |||
| XM_011545007.2 | XP_011543309.1 | 1 | 351 | |||
| XM_011545009.2 | XP_011543311.1 | 3 | 657 | |||
| XM_011545013.2 | XP_011543315.1 | 4 | 1120 | |||
| XM_011545014.2 | XP_011543316.1 | 4 | 1120 | |||
| XM_017017675.1 | XP_016873164.1 | 2 | 96 | |||
| XM_017017676.1 | XP_016873165.1 | 1 | 352 | |||
| XM_017017677.1 | XP_016873166.1 | 1 | 352 | |||
| XM_017017678.1 | XP_016873167.1 | 1 | 352 | |||
| XM_017017679.1 | XP_016873168.1 | 4 | 1120 | |||
| GenBank mRNA | AK128589.1 | - | 2 | 156 | ||
| AY138810.2 | - | 2 | 863 | |||
| AY325766.1 | - | 2 | 156 | |||

DNAJB13
Genomic Map