siRNA Details
| Gene Symbol: KRT77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175078.2 | NP_778253.2 | 4 | 920 | ||
| XM_011538288.2 | XP_011536590.1 | 4 | 682 | |||
| XM_011538289.2 | XP_011536591.1 | 4 | 981 | |||
| GenBank mRNA | AJ564104.1 | - | 4 | 920 | ||
| BC118598.1 | - | 5 | 1005 | 40108170 | ||
| BC122558.1 | - | 4 | 920 | 40108176 | ||

KRT77
Genomic Map