siRNA Details
| Gene Symbol: HDDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286451.1 | NP_001273380.1 | 3 | 288 | ||
| NM_198527.3 | NP_940929.1 | 3 | 288 | |||
| NR_104447.1 | 3 | 331 | ||||
| GenBank mRNA | AK057584.1 | - | 1 | 709 | ||
| BC033794.1 | - | 3 | 288 | 5122164 | ||
| BQ777217.1 | - | 3 | 238 | |||
| CV573934.1 | - | 3 | 273 | |||

HDDC3
Genomic Map