siRNA Details
Gene Symbol: C19orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198532.2 | NP_940934.1 | 3 | 392 | ||
GenBank mRNA | AK127680.1 | - | 3 | 386 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C19orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198532.2 | NP_940934.1 | 3 | 392 | ||
GenBank mRNA | AK127680.1 | - | 3 | 386 |