siRNA Details
| Gene Symbol: MROH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039464.3 | NP_001034553.2 | 3 | 774 | ||
| NR_026782.2 | 3 | 768 | ||||
| NR_111931.1 | 3 | 774 | ||||
| GenBank mRNA | AK093272.1 | - | 3 | 713 | ||
| AK130047.1 | - | 2 | 1263 | |||
| AK302446.1 | - | 3 | 721 | |||
| BC095478.1 | - | 2 | 556 | |||
| BC126126.1 | - | 1 | 474 | 8991842 | ||
| BC143891.1 | - | 1 | 474 | 9052408 | ||
| BC144516.1 | - | 3 | 712 | |||
| Gene Symbol: MROH7-TTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037639.2 | 3 | 879 | |||
| NR_037640.2 | 3 | 774 | ||||
| NR_037641.2 | 3 | 774 | ||||
| GenBank mRNA | BX648088.1 | - | 3 | 712 | ||
| CR749830.1 | - | 3 | 831 | |||

MROH7
Genomic Map