siRNA Details
| Gene Symbol: FAM89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198552.2 | NP_940954.1 | 2 | 579 | ||
| GenBank mRNA | BC009447.1 | - | 2 | 569 | 3530481 | |
| Gene Symbol: MIR1182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_031593.1 | 1 | 48 | |||

FAM89A
Genomic Map