siRNA Details
| Gene Symbol: TOPAZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145030.1 | NP_001138502.1 | 2 | 1341 | ||
| XM_011533694.2 | XP_011531996.1 | 2 | 1341 | |||
| XM_017006361.1 | XP_016861850.1 | 2 | 1341 | |||
| XM_017006362.1 | XP_016861851.1 | 2 | 1341 | |||
| GenBank mRNA | AK093476.1 | - | 2 | 1341 | ||

TOPAZ1
Genomic Map