siRNA Details
| Gene Symbol: VWC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198570.4 | NP_940972.2 | 4 | 1449 | ||
| NR_136188.1 | 3 | 996 | ||||
| GenBank mRNA | AB292671.1 | - | 3 | 893 | ||
| AY358393.1 | - | 4 | 1473 | |||
| BC110857.1 | - | 4 | 1392 | 5731863 | ||

VWC2
Genomic Map