siRNA Details
| Gene Symbol: NAT16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198571.2 | NP_940973.2 | 4 | 1333 | ||
| XM_005250325.1 | XP_005250382.1 | 5 | 1312 | |||
| XM_011516168.1 | XP_011514470.1 | 4 | 1294 | |||
| GenBank mRNA | AK096556.1 | - | 4 | 1333 | ||
| AL831911.1 | - | 2 | 603 | |||
| BC104841.1 | - | 5 | 1312 | 8143844 | ||

NAT16
Genomic Map