siRNA Details
| Gene Symbol: KCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135914.1 | NP_001129386.1 | 6 | 632 | ||
| NM_199349.2 | NP_955381.2 | 8 | 806 | |||
| XM_017012184.1 | XP_016867673.1 | 8 | 806 | |||
| XM_017012185.1 | XP_016867674.1 | 8 | 806 | |||
| XM_017012186.1 | XP_016867675.1 | 8 | 806 | |||
| XM_017012187.1 | XP_016867676.1 | 8 | 806 | |||
| XM_017012188.1 | XP_016867677.1 | 8 | 806 | |||
| XM_017012189.1 | XP_016867678.1 | 8 | 806 | |||
| XM_017012190.1 | XP_016867679.1 | 8 | 806 | |||
| XM_017012191.1 | XP_016867680.1 | 2 | 105 | |||
| XM_017012192.1 | XP_016867681.1 | 2 | 116 | |||
| XM_017012193.1 | XP_016867682.1 | 8 | 806 | |||
| XM_017012194.1 | XP_016867683.1 | 8 | 806 | |||
| GenBank mRNA | AK095464.1 | - | 2 | 204 | ||
| AK122706.1 | - | 8 | 806 | |||

KCP
Genomic Map