siRNA Details
| Gene Symbol: CATSPER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198137.1 | NP_937770.1 | 2, 3 | 365 | ||
| XM_011541432.2 | XP_011539734.1 | 2, 3 | 472 | |||
| XM_011541433.2 | XP_011539735.1 | 2, 3 | 1031 | |||
| GenBank mRNA | BC128138.1 | - | 2, 3 | 364 | 40112485 | |
| BC128139.1 | - | 2, 3 | 365 | 40112493 | ||
| BC144663.1 | - | 2, 3 | 432 | |||

CATSPER4
Genomic Map