siRNA Details
| Gene Symbol: KEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000420.2 | NP_000411.1 | 4, 5 | 608 | ||
| XM_005249993.2 | XP_005250050.1 | 4, 5 | 459 | |||
| GenBank mRNA | AK314831.1 | - | 4, 5 | 509 | ||
| BC003135.1 | - | 4, 5 | 608 | 2906155 | ||
| BC050639.1 | - | 4, 5 | 503 | 5742350 | ||
| FN998902.1 | - | 3, 4 | 398 | |||
| HE572750.1 | - | 3, 4 | 398 | |||
| M64934.2 | - | 4, 5 | 518 | |||

KEL
Genomic Map